Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304777.1 | NP_001291706.1 | 7 | 2439 | |
NM_001304779.1 | NP_001291708.1 | 4 | 2078 | ||
NM_016041.4 | NP_057125.2 | 7 | 2442 | ||
NR_130905.1 | 6 | 2554 | |||
NR_130906.1 | 6 | 2348 | |||
GenBank mRNA | AK092218.1 | - | 1 | 208 |