Gene Transcripts

Gene Symbol
WDR86
Entrez Gene ID
349136
Gene Name
WD repeat domain 86
Gene Aliases
Chromosome Location
Chr. 7: 151377974 - 151410705 on Build GRCh38
UniGene ID
Hs.647083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006715966.2 XP_006716029.1 5 1821
GenBank mRNA AK096225.1 - 1 784

View Full Product Details