Gene Transcripts
- Gene Symbol
- MAGI2-AS3
- Entrez Gene ID
- 100505881
- Gene Name
- MAGI2 antisense RNA 3
- Gene Aliases
- Chromosome Location
- Chr. 7: 79452957 - 79471208 on Build GRCh38
- UniGene ID
- Hs.31474
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR749529.1 |
- |
1 |
77 |
|
View Full Product Details