Gene Transcripts
- Gene Symbol
- HHIPL1
- Entrez Gene ID
- 84439
- Gene Name
- HHIP like 1
- Gene Aliases
- KIAA1822, UNQ9245
- Chromosome Location
- Chr. 14: 99604476 - 99680569 on Build GRCh38
- UniGene ID
- Hs.131714
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC039687.1 |
- |
2 |
722 |
|
View Full Product Details