Gene Transcripts
- Gene Symbol
- HNF4A-AS1
- Entrez Gene ID
- 101927219
- Gene Name
- HNF4A antisense RNA 1
- Gene Aliases
- uc002xlx
- Chromosome Location
- Chr. 20: 44377743 - 44395706 on Build GRCh38
- UniGene ID
- Hs.655919
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109949.1 |
|
3 |
452 |
|
GenBank mRNA |
AF143870.1 |
- |
3 |
463 |
|
BC071794.1 |
- |
3 |
655 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000452481.1 |
- |
3 |
452 |
|
Chr. 20 - 44377743 - 44395706 [-] |
View Full Product Details