Gene Transcripts
- Gene Symbol
- C3orf38
- Entrez Gene ID
- 285237
- Gene Name
- chromosome 3 open reading frame 38
- Gene Aliases
- Chromosome Location
- Chr. 3: 88149713 - 88157965 on Build GRCh38
- UniGene ID
- Hs.476944
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016013.1 |
- |
1 |
1312 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
D45548.1 |
- |
1 |
176 |
|
Chr. 3 - 88166233 - 88166469 [+] |
View Full Product Details