Gene Transcripts
- Gene Symbol
- HIP1
- Entrez Gene ID
- 3092
- Gene Name
- huntingtin interacting protein 1
- Gene Aliases
- HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
- Chromosome Location
- Chr. 7: 75533298 - 75738976 on Build GRCh38
- UniGene ID
- Hs.329266
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY358103.1 |
- |
1 |
321 |
|
JX965370.1 |
- |
2 |
489 |
|
JX965371.1 |
- |
2 |
371 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR745472.1 |
- |
2 |
489 |
|
Chr. 7 - 75733460 - 75738962 [-] |
GenBankmRNA |
BF998227.1 |
- |
1 |
157 |
|
Chr. 7 - 75733416 - 75733630 [-] |
View Full Product Details