Gene Transcripts
- Gene Symbol
- INVS
- Entrez Gene ID
- 27130
- Gene Name
- inversin
- Gene Aliases
- INV, NPH2, NPHP2
- Chromosome Location
- Chr. 9: 100099185 - 100302175 on Build GRCh38
- UniGene ID
- Hs.558477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC110864.1 |
- |
3 |
1179 |
|
View Full Product Details