Gene Transcripts
- Gene Symbol
- RAB11FIP3
- Entrez Gene ID
- 9727
- Gene Name
- RAB11 family interacting protein 3
- Gene Aliases
- CART1, Rab11-FIP3
- Chromosome Location
- Chr. 16: 425621 - 522485 on Build GRCh38
- UniGene ID
- Hs.531642
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE799799.1 |
- |
1 |
69 |
|
Chr. 16 - 497340 - 497978 [+] |
GenBankmRNA |
AK094817.1 |
- |
1 |
2216 |
|
Chr. 16 - 495153 - 497939 [+] |
View Full Product Details