Gene Transcripts
- Gene Symbol
- PSPH
- Entrez Gene ID
- 5723
- Gene Name
- phosphoserine phosphatase
- Gene Aliases
- PSP, PSPHD
- Chromosome Location
- Chr. 7: 56010363 - 56051604 on Build GRCh38
- UniGene ID
- Hs.512656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054677.1 |
- |
5 |
1709 |
|
BC052958.1 |
- |
3 |
2639 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ424231.1 |
- |
1 |
638 |
|
Chr. 7 - 56040373 - 56041201 [-] |
View Full Product Details