Gene Transcripts

Gene Symbol
SYT16
Entrez Gene ID
83851
Gene Name
synaptotagmin 16
Gene Aliases
CHR14SYT, SYT14L, Strep14, syt14r, yt14r
Chromosome Location
Chr. 14: 61812156 - 62112825 on Build GRCh38
UniGene ID
Hs.404139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006720272.3 XP_006720335.1 8 14228
XM_011537222.2 XP_011535524.1 8 14136
XM_011537223.2 XP_011535525.1 7 13342
XM_011537224.2 XP_011535526.1 8 13488
XM_011537225.2 XP_011535527.1 8 13787
XM_011537227.2 XP_011535529.2 6 13165
XM_011537228.2 XP_011535530.1 6 13108
XM_011537229.2 XP_011535531.1 5 12952
XM_017021693.1 XP_016877182.1 9 4904
XM_017021694.1 XP_016877183.1 8 13994
XM_017021695.1 XP_016877184.1 9 14139
XM_017021696.1 XP_016877185.1 7 13287
XM_017021697.1 XP_016877186.1 7 13126
XM_017021698.1 XP_016877187.1 7 13411
GenBank mRNA AK055647.1 - 1 1466

View Full Product Details