Gene Transcripts
- Gene Symbol
- LOC102724765
- Entrez Gene ID
- 102724765
- Gene Name
- uncharacterized LOC102724765
- Gene Aliases
- Chromosome Location
- Chr. 6: 25014952 - 25036447 on Build GRCh38
- UniGene ID
- Hs.737906
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC070382.1 |
- |
1 |
59 |
|
- Gene Symbol
- FAM65B
- Entrez Gene ID
- 9750
- Gene Name
- family with sequence similarity 65 member B
- Gene Aliases
- C6orf32, DFNB104, DIFF40, DIFF48, MYONAP, PL48
- Chromosome Location
- Chr. 6: 24804281 - 25042288 on Build GRCh38
- UniGene ID
- Hs.559459
- Species
- Homo sapiens
View Full Product Details