Gene Transcripts

Gene Symbol
WWTR1
Entrez Gene ID
25937
Gene Name
WW domain containing transcription regulator 1
Gene Aliases
TAZ
Chromosome Location
Chr. 3: 149517235 - 149724783 on Build GRCh38
UniGene ID
Hs.477921
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127620.1 - 1 2318


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BP390394.1 - 1 386 Chr. 3 - 149571237 - 149571703 [-]

View Full Product Details