Gene Transcripts
- Gene Symbol
- TRIM13
- Entrez Gene ID
- 10206
- Gene Name
- tripartite motif containing 13
- Gene Aliases
- CAR, DLEU5, LEU5, RFP2, RNF77
- Chromosome Location
- Chr. 13: 49997007 - 50018467 on Build GRCh38
- UniGene ID
- Hs.436922
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC029514.1 |
- |
1 |
1356 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF279660.3 |
- |
1 |
1547 |
|
Chr. 13 - 49996975 - 50344526 [+] |
View Full Product Details