Gene Transcripts

Gene Symbol
SSTR3
Entrez Gene ID
6753
Gene Name
somatostatin receptor 3
Gene Aliases
SS-3-R, SS3-R, SS3R, SSR-28
Chromosome Location
Chr. 22: 37204237 - 37220451 on Build GRCh38
UniGene ID
Hs.225995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BF514979.1 - 1 355


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY273569.1 - 1 369 Chr. 22 - 37204232 - 37204653 [+]
GenBankmRNA BC133662.1 - 1 63 Chr. 22 - 37204521 - 37205449 [+]

View Full Product Details