Gene Transcripts
- Gene Symbol
- HSPB8
- Entrez Gene ID
- 26353
- Gene Name
- heat shock protein family B (small) member 8
- Gene Aliases
- CMT2L, DHMN2, E2IG1, H11, HMN2, HMN2A, HSP22
- Chromosome Location
- Chr. 12: 119178790 - 119194746 on Build GRCh38
- UniGene ID
- Hs.400095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001825.1 |
- |
1 |
1298 |
|
View Full Product Details