Gene Transcripts

Gene Symbol
PDCD6IP
Entrez Gene ID
10015
Gene Name
programmed cell death 6 interacting protein
Gene Aliases
AIP1, ALIX, DRIP4, HP95
Chromosome Location
Chr. 3: 33798571 - 33869707 on Build GRCh38
UniGene ID
Hs.475896
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BQ895877.1 - 1 627


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR622191.1 - 1 886 Chr. 3 - 33797188 - 33799063 [-]

View Full Product Details