Gene Transcripts
- Gene Symbol
- DNM1
- Entrez Gene ID
- 1759
- Gene Name
- dynamin 1
- Gene Aliases
- DNM, EIEE31
- Chromosome Location
- Chr. 9: 128203355 - 128255248 on Build GRCh38
- UniGene ID
- Hs.522413
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127038.1 |
- |
1 |
995 |
|
View Full Product Details