Gene Transcripts
- Gene Symbol
- TTC12
- Entrez Gene ID
- 54970
- Gene Name
- tetratricopeptide repeat domain 12
- Gene Aliases
- TPARM
- Chromosome Location
- Chr. 11: 113314529 - 113385952 on Build GRCh38
- UniGene ID
- Hs.288772
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125909.1 |
- |
1 |
2714 |
|
View Full Product Details