Gene Transcripts

Gene Symbol
Gprin2
Entrez Gene ID
432839
Gene Name
G protein regulated inducer of neurite outgrowth 2
Gene Aliases
C130040D06Rik, C230073P13, Gm286, mKIAA0514
Chromosome Location
Chr. 14: 34185688 - 34201692 on Build GRCm38
UniGene ID
Mm.294643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK048200.1 - 1 995
AK082455.1 - 1 258


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CX730911.1 - 1 362 Chr. 14 - 34186136 - 34186770 [-]
GenBankmRNA BB651025.1 - 1 256 Chr. 14 - 34185987 - 34186664 [-]
GenBankmRNA BB367967.2 - 1 344 Chr. 14 - 34186104 - 34186741 [-]

View Full Product Details