Gene Transcripts
- Gene Symbol
- Osr2
- Entrez Gene ID
- 107587
- Gene Name
- odd-skipped related 2
- Gene Aliases
- 5430409I15Rik, Osr2A, Osr2B
- Chromosome Location
- Chr. 15: 35296112 - 35303308 on Build GRCm38
- UniGene ID
- Mm.46336
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017286.1 |
- |
2 |
1123 |
|
View Full Product Details