Gene Transcripts
- Gene Symbol
- Enpp1
- Entrez Gene ID
- 18605
- Gene Name
- ectonucleotide pyrophosphatase/phosphodiesterase 1
- Gene Aliases
- 4833416E15Rik, AI428932, C76301, CD203c, E-NPP 1, E-NPP1, Ly-41, M6S1, NPP1, Npps, PC-1, Pca, Pca-1, Pdnp1, ttw, twy
- Chromosome Location
- Chr. 10: 24637914 - 24712154 on Build GRCm38
- UniGene ID
- Mm.27254
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014707.1 |
- |
1 |
837 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CJ257496.1 |
- |
1 |
487 |
|
Chr. 10 - 24637920 - 24638466 [-] |
View Full Product Details