Gene Transcripts
- Gene Symbol
- Tctn3
- Entrez Gene ID
- 67590
- Gene Name
- tectonic family member 3
- Gene Aliases
- 4930521E07Rik, AI197391, Tect3
- Chromosome Location
- Chr. 19: 40596446 - 40612281 on Build GRCm38
- UniGene ID
- Mm.374056
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK155612.1 |
- |
|
195 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY749708.1 |
- |
4 |
195 |
|
Chr. 19 - 40589435 - 40594419 [+] |
View Full Product Details