Gene Transcripts

Gene Symbol
Mir99ahg
Entrez Gene ID
77994
Gene Name
Mir99a and Mirlet7c-1 host gene (non-protein coding)
Gene Aliases
2810055G20Rik, D730043B02Rik
Chromosome Location
Chr. 16: 77329328 - 77558428 on Build GRCm38
UniGene ID
Mm.394688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK033222.1 - 698
AK082961.1 - 1198


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB649986.1 - 1 124 Chr. 16 - 77594902 - 77773427 [+]

View Full Product Details