Gene Transcripts
- Gene Symbol
- Sntg1
- Entrez Gene ID
- 71096
- Gene Name
- syntrophin, gamma 1
- Gene Aliases
- 4933426D16Rik, G1SYN, SYN4
- Chromosome Location
- Chr. 1: 8359739 - 9301061 on Build GRCm38
- UniGene ID
- Mm.336395
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080155.1 |
- |
5 |
1579 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB221905.2 |
- |
5 |
539 |
|
Chr. 1 - 8950572 - 9298236 [-] |
View Full Product Details