Gene Transcripts
- Gene Symbol
- Zeb1
- Entrez Gene ID
- 21417
- Gene Name
- zinc finger E-box binding homeobox 1
- Gene Aliases
- 3110032K11Rik, AREB6, BZP, MEB1, Nil2, TCF-8, Tcf18, Tcf8, Tw, ZEB, Zfhep, Zfhx1a, Zfx1a, Zfx1ha, [delta]EF1
- Chromosome Location
- Chr. 18: 5591860 - 5775468 on Build GRCm38
- UniGene ID
- Mm.3929
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK041408.1 |
- |
1, 2 |
348 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB638079.1 |
- |
1, 2 |
348 |
|
Chr. 18 - 5603744 - 5641904 [+] |
View Full Product Details