Gene Transcripts
- Gene Symbol
- Adh1
- Entrez Gene ID
- 11522
- Gene Name
- alcohol dehydrogenase 1 (class I)
- Gene Aliases
- ADH-AA, AI194826, Adh-1, Adh-1-t, Adh-1e, Adh-1t, Adh-3e, Adh1-e, Adh1-t, Adh1tl, Adh3-e
- Chromosome Location
- Chr. 3: 138277645 - 138290691 on Build GRCm38
- UniGene ID
- Mm.2409
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082149.1 |
- |
|
393 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB650573.1 |
- |
|
403 |
|
Chr. - - [-] |
View Full Product Details