Gene Transcripts
- Gene Symbol
- Npy1r
- Entrez Gene ID
- 18166
- Gene Name
- neuropeptide Y receptor Y1
- Gene Aliases
- Npyr, Y1-R
- Chromosome Location
- Chr. 8: 66697192 - 66706798 on Build GRCm38
- UniGene ID
- Mm.5112
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK162898.1 |
- |
3 |
464 |
|
View Full Product Details