Gene Transcripts
- Gene Symbol
- Hcfc2
- Entrez Gene ID
- 67933
- Gene Name
- host cell factor C2
- Gene Aliases
- 1700129L13Rik, AU019838
- Chromosome Location
- Chr. 10: 82697497 - 82795865 on Build GRCm38
- UniGene ID
- Mm.127405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018461.1 |
- |
1 |
1848 |
|
AK040565.1 |
- |
1 |
1855 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CX768124.1 |
- |
1 |
495 |
|
Chr. 10 - 82740590 - 82741256 [+] |
GenBankmRNA |
CF535355.1 |
- |
1 |
136 |
|
Chr. 10 - 82740948 - 82741395 [+] |
View Full Product Details