Gene Transcripts
- Gene Symbol
- Skint1
- Entrez Gene ID
- 639781
- Gene Name
- selection and upkeep of intraepithelial T cells 1
- Gene Aliases
- OTTMUSG00000008542, Skint-1
- Chromosome Location
- Chr. 4: 112006269 - 112029538 on Build GRCm38
- UniGene ID
- Mm.398873
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029209.1 |
- |
2 |
578 |
|
View Full Product Details