Gene Transcripts
- Gene Symbol
- Psen1
- Entrez Gene ID
- 19164
- Gene Name
- presenilin 1
- Gene Aliases
- Ad3h, PS-1, PS1, S182
- Chromosome Location
- Chr. 12: 83688146 - 83738446 on Build GRCm38
- UniGene ID
- Mm.998
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK148390.1 |
- |
1 |
1492 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB771315.1 |
- |
1 |
466 |
|
Chr. 12 - 83737851 - 83738370 [+] |
View Full Product Details