Gene Transcripts
- Gene Symbol
- Tanc2
- Entrez Gene ID
- 77097
- Gene Name
- tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
- Gene Aliases
- 3526402J09Rik, 5730590C14Rik
- Chromosome Location
- Chr. 11: 105589986 - 105929304 on Build GRCm38
- UniGene ID
- Mm.22501
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019982.1 |
- |
1 |
785 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DV064943.1 |
- |
|
286 |
|
Chr. - - [-] |
View Full Product Details