Gene Transcripts

Gene Symbol
Zzz3
Entrez Gene ID
108946
Gene Name
zinc finger, ZZ domain containing 3
Gene Aliases
3110065C23Rik, 6430567E01Rik, AA408566, AI481057, AW556059, R75514
Chromosome Location
Chr. 3: 152394486 - 152462826 on Build GRCm38
UniGene ID
Mm.379292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK165882.1 - 1 50


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY734015.1 - 1 47 Chr. 3 - 152395475 - 152448910 [+]

View Full Product Details