Gene Transcripts
- Gene Symbol
- Asah1
- Entrez Gene ID
- 11886
- Gene Name
- N-acylsphingosine amidohydrolase 1
- Gene Aliases
- 2310081N20Rik, AC, Asah
- Chromosome Location
- Chr. 8: 41340643 - 41374697 on Build GRCm38
- UniGene ID
- Mm.22547
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034204.1 |
- |
|
3225 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB080497.2 |
- |
1 |
449 |
|
Chr. 8 - 41339838 - 41340507 [-] |
View Full Product Details