Gene Transcripts
- Gene Symbol
- Dnajc1
- Entrez Gene ID
- 13418
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 1
- Gene Aliases
- 4733401K02Rik, AA960110, D230036H06Rik, Dnajl1, ERdj1, ERj1p, MTJ1
- Chromosome Location
- Chr. 2: 18206333 - 18392851 on Build GRCm38
- UniGene ID
- Mm.246674
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014639.1 |
- |
1 |
249 |
|
View Full Product Details