Gene Transcripts

Gene Symbol
Otud3
Entrez Gene ID
73162
Gene Name
OTU domain containing 3
Gene Aliases
3110030K17Rik
Chromosome Location
Chr. 4: 138891063 - 138913947 on Build GRCm38
UniGene ID
Mm.238081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK076876.1 - 1 894


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AI036198.1 - 1 217 Chr. 4 - 138891071 - 138891447 [-]
GenBankmRNA AV267369.2 - 1 487 Chr. 4 - 138891072 - 138891721 [-]
GenBankmRNA BB706496.1 - 1 177 Chr. 4 - 138891070 - 138891415 [-]
GenBankmRNA BB810207.1 - 1 242 Chr. 4 - 138891066 - 138891480 [-]

View Full Product Details