Gene Transcripts
- Gene Symbol
- Mettl1
- Entrez Gene ID
- 17299
- Gene Name
- methyltransferase like 1
- Gene Aliases
- 2810012D02Rik
- Chromosome Location
- Chr. 10: 127041643 - 127046365 on Build GRCm38
- UniGene ID
- Mm.251593
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012721.1 |
- |
1 |
380 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CA547711.1 |
- |
1 |
321 |
|
Chr. 10 - 127045931 - 127046439 [+] |
GenBankmRNA |
BY711843.1 |
- |
1 |
380 |
|
Chr. 10 - 127045871 - 127046363 [+] |
GenBankmRNA |
CJ303604.1 |
- |
1 |
290 |
|
Chr. 10 - 127045961 - 127046364 [+] |
View Full Product Details