Gene Transcripts
- Gene Symbol
- Znrf2
- Entrez Gene ID
- 387524
- Gene Name
- zinc and ring finger 2
- Gene Aliases
- 1190002C14Rik, AU044205, D6Ertd365e
- Chromosome Location
- Chr. 6: 54816916 - 54890224 on Build GRCm38
- UniGene ID
- Mm.286149
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK078666.1 |
- |
|
1680 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA790403.1 |
- |
1 |
461 |
|
Chr. 6 - 54890369 - 54890892 [+] |
GenBankmRNA |
CX729999.1 |
- |
1 |
559 |
|
Chr. 6 - 54890273 - 54890891 [+] |
GenBankmRNA |
BU699828.1 |
- |
1 |
460 |
|
Chr. 6 - 54890372 - 54890896 [+] |
GenBankmRNA |
BB324362.2 |
- |
1 |
523 |
|
Chr. 6 - 54890312 - 54890950 [+] |
View Full Product Details