Gene Transcripts
- Gene Symbol
- Krtap6-3
- Entrez Gene ID
- 100040249
- Gene Name
- keratin associated protein 6-3
- Gene Aliases
- HGTpII.3
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.328678
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
D89901.1 |
- |
|
191 |
|
View Full Product Details