Gene Transcripts
- Gene Symbol
- Nova2
- Entrez Gene ID
- 384569
- Gene Name
- neuro-oncological ventral antigen 2
- Gene Aliases
- Gm1424
- Chromosome Location
- Chr. 7: 18925888 - 18965319 on Build GRCm38
- UniGene ID
- Mm.262724
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC062110.1 |
- |
1 |
685 |
|
View Full Product Details