Gene Transcripts
- Gene Symbol
- Mirg
- Entrez Gene ID
- 100040724
- Gene Name
- miRNA containing gene
- Gene Aliases
- 2810474H01Rik, AI852992, Gm2928, Meg9, mir-410
- Chromosome Location
- Chr. 12: 109734981 - 109749458 on Build GRCm38
- UniGene ID
- Mm.260543
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077315.1 |
- |
2 |
1587 |
|
View Full Product Details