Gene Transcripts
- Gene Symbol
- Tmppe
- Entrez Gene ID
- 100504715
- Gene Name
- transmembrane protein with metallophosphoesterase domain
- Gene Aliases
- Chromosome Location
- Chr. 9: 114401095 - 114411201 on Build GRCm38
- UniGene ID
- Mm.290516
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082033.1 |
- |
1 |
696 |
|
View Full Product Details