Gene Transcripts

Gene Symbol
Tmppe
Entrez Gene ID
100504715
Gene Name
transmembrane protein with metallophosphoesterase domain
Gene Aliases
Chromosome Location
Chr. 9: 114401095 - 114411201 on Build GRCm38
UniGene ID
Mm.290516
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK082033.1 - 1 484


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU698502.1 - 1 323 Chr. 9 - 114409812 - 114410306 [+]
GenBankmRNA BU695488.1 - 1 323 Chr. 9 - 114409812 - 114410319 [+]
GenBankmRNA BU696771.1 - 1 323 Chr. 9 - 114409812 - 114410319 [+]
GenBankmRNA BU694508.1 - 1 323 Chr. 9 - 114409812 - 114410345 [+]

View Full Product Details