Gene Transcripts
- Gene Symbol
- Mib1
- Entrez Gene ID
- 225164
- Gene Name
- mindbomb homolog 1 (Drosophila)
- Gene Aliases
- DIP-1, E430019M12Rik, Mib, mKIAA1323
- Chromosome Location
- Chr. 18: 10725625 - 10818578 on Build GRCm38
- UniGene ID
- Mm.21500
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CX209849.1 |
- |
1 |
375 |
|
Chr. 18 - 10816366 - 10817117 [+] |
GenBankmRNA |
BU055391.1 |
- |
1 |
267 |
|
Chr. 18 - 10816463 - 10817166 [+] |
GenBankmRNA |
AA178681.1 |
- |
1 |
164 |
|
Chr. 18 - 10816564 - 10817026 [+] |
View Full Product Details