Gene Transcripts

Gene Symbol
Crnde
Entrez Gene ID
71296
Gene Name
colorectal neoplasia differentially expressed (non-protein coding)
Gene Aliases
4933436C20Rik
Chromosome Location
Chr. 8: 92326031 - 92356120 on Build GRCm38
UniGene ID
Mm.86664
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033641.3 7 1155
NR_110452.1 4 974
GenBank mRNA AK017076.1 - 7 1155
CX205694.1 - 4 685

View Full Product Details