Gene Transcripts
- Gene Symbol
- Myt1l
- Entrez Gene ID
- 17933
- Gene Name
- myelin transcription factor 1-like
- Gene Aliases
- 2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106
- Chromosome Location
- Chr. 12: 29528311 - 29923216 on Build GRCm38
- UniGene ID
- Mm.253067
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139366.1 |
- |
1 |
654 |
|
BC056457.1 |
- |
|
743 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB267486.2 |
- |
1 |
648 |
|
Chr. 12 - 29528367 - 29529024 [+] |
View Full Product Details