Gene Transcripts
- Gene Symbol
- Enah
- Entrez Gene ID
- 13800
- Gene Name
- enabled homolog (Drosophila)
- Gene Aliases
- Mena, NDPP-1, Ndpp1, WBP8
- Chromosome Location
- Chr. 1: 181896384 - 182019980 on Build GRCm38
- UniGene ID
- Mm.389224
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK081526.1 |
- |
1 |
486 |
|
View Full Product Details