Gene Transcripts

Gene Symbol
Eny2
Entrez Gene ID
223527
Gene Name
enhancer of yellow 2 homolog (Drosophila)
Gene Aliases
1810057B09Rik, 6720481I12, DC6, Ey2
Chromosome Location
Chr. 15: 44428081 - 44438256 on Build GRCm38
UniGene ID
Mm.291828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK016562.1 - 1 890
AK087816.1 - 1 892


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB556629.2 - 1 353 Chr. 15 - 44428684 - 44429347 [+]

View Full Product Details