Gene Transcripts
- Gene Symbol
- Eny2
- Entrez Gene ID
- 223527
- Gene Name
- enhancer of yellow 2 homolog (Drosophila)
- Gene Aliases
- 1810057B09Rik, 6720481I12, DC6, Ey2
- Chromosome Location
- Chr. 15: 44428081 - 44438256 on Build GRCm38
- UniGene ID
- Mm.291828
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016562.1 |
- |
1 |
506 |
|
AK087816.1 |
- |
1 |
506 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU518875.1 |
- |
1 |
61 |
|
Chr. 15 - 44428591 - 44429405 [+] |
View Full Product Details