Gene Transcripts
- Gene Symbol
- Nvl
- Entrez Gene ID
- 67459
- Gene Name
- nuclear VCP-like
- Gene Aliases
- 1200009I24Rik
- Chromosome Location
- Chr. 1: 181087140 - 181144215 on Build GRCm38
- UniGene ID
- Mm.263464
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK134686.1 |
- |
1 |
289 |
|
AK145078.1 |
- |
1 |
2836 |
|
View Full Product Details